Methyl-6-brom-7-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7112-1G
COBBPY-7112-1GEA 310 EUR
COBBPY-7112-1G
Methyl-6-brom-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Methyl-6-brom-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₁BrN₂O₂
MDL: MFCD09475851
CAS-Nummer: 907945-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR