Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7113-1G
COBBPY-7113-1GEA 0 EUR
COBBPY-7113-1G
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₁BrN₂O₂
MDL: MFCD09475853
CAS-Nummer: 135995-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR