Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7114-1G
COBBPY-7114-1GEA 149 EUR
COBBPY-7114-1G
Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₁BrN₂O₂
MDL: MFCD09475852
CAS-Nummer: 847446-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR