6-Brom-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridin
Lieferant: COMBI-BLOCKS
PY-7141-1G
COBBPY-7141-1GEA
690
EUR
COBBPY-7141-1G
6-Brom-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridin
6-Brom-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridin
Formel:
C₇H₆BrN₃S |
MDL:
MFCD09475863 CAS-Nummer: 941294-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...