Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7142-1G PY-7142-5G
COBBPY-7142-1GEA 0 EUR
COBBPY-7142-1G COBBPY-7142-5G
Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₈BrClN₂O₂
MDL: MFCD09607953
CAS-Nummer: 951884-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR