Methyl-5-brom-2-chlornicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 5-bromo-2-chloronicotinate

PY-7148-5G
COBBPY-7148-5GEA 48 EUR
COBBPY-7148-5G
Methyl-5-brom-2-chlornicotinat
Methyl-5-brom-2-chlornicotinat
Formel: C₇H₅BrClNO₂
Molecular Weight: 250,48 g/mol
MDL: MFCD03844848
CAS-Nummer: 78686-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR