6,7-Dichlor-2-methyl-3-(trifluormethyl)chinoxalin

Lieferant: COMBI-BLOCKS

PY-7180-1G
COBBPY-7180-1GEA 0 EUR
COBBPY-7180-1G
6,7-Dichlor-2-methyl-3-(trifluormethyl)chinoxalin
6,7-Dichloro-2-methyl-3-(trifluoromethyl)quinoxaline
Formel: C₁₀H₅Cl₂F₃N₂
MDL: MFCD09801054
CAS-Nummer: 143309-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR