Ethyl-6-chlorpicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

PY-7261-1G PY-7261-5G
COBBPY-7261-1GEA 38.2 EUR
COBBPY-7261-1G COBBPY-7261-5G
Ethyl-6-chlorpicolinat
Ethyl-6-chlorpicolinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185.61 g/mol
MDL: MFCD09878438
CAS-Nummer: 21190-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR