5-Brom-2-chlor-4-iodpyridin
Lieferant: COMBI-BLOCKS
PY-7284-1G
PY-7284-5G
COBBPY-7284-1GEA
227
EUR
COBBPY-7284-1G
COBBPY-7284-5G
5-Brom-2-chlor-4-iodpyridin
5-Brom-2-chlor-4-iodpyridin
Formel:
C₅H₂BrClIN Molecular Weight: 318.34 g/mol |
MDL:
MFCD09972189 CAS-Nummer: 401892-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...