2-Methoxy-5-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

PY-7303-100G PY-7303-25G
COBBPY-7303-100GEA 124 EUR
COBBPY-7303-100G COBBPY-7303-25G
2-Methoxy-5-nitropyridin
2-Methoxy-5-nitropyridine
Formel: C₆H₆N₂O₃
Molecular Weight: 154.13 g/mol
Schmelzpunkt: 114
MDL: MFCD00006263
CAS-Nummer: 5446-92-4
EINECS: 226-661-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR