5-Brom-3-nitro-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

PY-7307-1G PY-7307-5G
COBBPY-7307-1GEA 126 EUR
COBBPY-7307-1G COBBPY-7307-5G
5-Brom-3-nitro-2-(4-morpholino)pyridin
5-Brom-3-nitro-2-(4-morpholino)pyridin
Formel: C₉H₁₀BrN₃O₃
MDL: MFCD02736520
CAS-Nummer: 505052-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR