5-Brom-2-chlor-4-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7317-5G PY-7317-1G
COBBPY-7317-5GEA 127 EUR
COBBPY-7317-5G COBBPY-7317-1G
5-Brom-2-chlor-4-methyl-3-nitropyridin
5-Brom-2-chlor-4-methyl-3-nitropyridin
Formel: C₆H₄BrClN₂O₂
Molecular Weight: 251.47 g/mol
MDL: MFCD06659505
CAS-Nummer: 884495-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR