2-(Cyclopentylamino)-3-nitro-5-picolin

Lieferant: COMBI-BLOCKS

PY-7319-1G PY-7319-5G
COBBPY-7319-1GEA 49.7 EUR
COBBPY-7319-1G COBBPY-7319-5G
2-(Cyclopentylamino)-3-nitro-5-picolin
2-(Cyclopentylamino)-3-nitro-5-picolin
Formel: C₁₁H₁₅N₃O₂
MDL: MFCD10699704
CAS-Nummer: 1033202-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR