3-Fluor-2-hydroxypyridin
Lieferant: COMBI-BLOCKS
PY-7344-1G
PY-7344-25G
PY-7344-5G
COBBPY-7344-1GEA
35.9
EUR
COBBPY-7344-1G
COBBPY-7344-25G
COBBPY-7344-5G
3-Fluor-2-hydroxypyridin
3-Fluor-2-hydroxypyridin
Formel:
C₅H₄FNO Molecular Weight: 113.09 g/mol |
MDL:
MFCD03095042 CAS-Nummer: 1547-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...