3,3'-Bipyridin-5,5'-dicarbonitril

Lieferant: COMBI-BLOCKS

PY-7346-1G
COBBPY-7346-1GEA 204 EUR
COBBPY-7346-1G
3,3'-Bipyridin-5,5'-dicarbonitril
3,3'-Bipyridin-5,5'-dicarbonitril
Formel: C₁₂H₆N₄
MDL: MFCD16295131
CAS-Nummer: 1226808-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR