3-Brom-2-chlor-5-methoxypyridin

Lieferant: COMBI-BLOCKS

PY-7351-1G
COBBPY-7351-1GEA 742 EUR
COBBPY-7351-1G
3-Brom-2-chlor-5-methoxypyridin
3-Bromo-2-chloro-5-methoxypyridine
Formel: C₆H₅BrClNO
MDL: MFCD10699708
CAS-Nummer: 1033202-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR