5-Methyl-2-pyrazincarbonsäure

Lieferant: COMBI-BLOCKS

PY-7724-100G PY-7724-25G
COBBPY-7724-100GEA 187 EUR
COBBPY-7724-100G COBBPY-7724-25G
5-Methyl-2-pyrazincarbonsäure
5-Methylpyrazine-2-carboxylic acid
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
MDL: MFCD00068241
CAS-Nummer: 5521-55-1
EINECS: 413-260-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR