Methyl-6-formylpicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-formylpyridine-2-carboxylate, Methyl 6-formyl-2-pyridinecarboxylate

PY-7741-1G
COBBPY-7741-1GEA 125 EUR
COBBPY-7741-1G
Methyl-6-formylpicolinat
Methyl-6-formylpicolinat
Formel: C₈H₇NO₃
Molecular Weight: 165,1500 g/mol
MDL: MFCD07367925
CAS-Nummer: 69950-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR