2-Amino-5-chlor-3-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 5-Chlor-3-nitropyridin-2-amin, 5-Chlor-3-nitro-2-pyridylamin

PY-7763-25G PY-7763-5G
COBBPY-7763-25GEA 76.1 EUR
COBBPY-7763-25G COBBPY-7763-5G
2-Amino-5-chlor-3-nitropyridin
2-Amino-5-chlor-3-nitropyridin
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173.56 g/mol
MDL: MFCD00092011
CAS-Nummer: 5409-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR