Methyl-2-methoxynicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-methoxynicotinate

PY-7832-25G
COBBPY-7832-25GEA 122 EUR
COBBPY-7832-25G
Methyl-2-methoxynicotinat
Methyl-2-methoxynicotinat
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD06801052
CAS-Nummer: 67367-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR