2-Chlor-6-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chlor-5-nitro-2-picolin, 6-Chlor-2-methyl-5-nitropyridin, 2-Chlor-3-nitro-6-picolin

PY-7876-1G PY-7876-5G
COBBPY-7876-1GEA 0 EUR
COBBPY-7876-1G COBBPY-7876-5G
2-Chlor-6-methyl-3-nitropyridin
2-Chloro-6-methyl-3-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.57 g/mol
MDL: MFCD03085820
CAS-Nummer: 56057-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR