2-Chlor-6-methyl-3-nitropyridin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Chlor-5-nitro-2-picolin, 6-Chlor-2-methyl-5-nitropyridin, 2-Chlor-3-nitro-6-picolin
PY-7876-1G
PY-7876-5G
COBBPY-7876-1GEA
0
EUR
COBBPY-7876-1G
COBBPY-7876-5G
2-Chlor-6-methyl-3-nitropyridin
2-Chloro-6-methyl-3-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172.57 g/mol |
MDL:
MFCD03085820 CAS-Nummer: 56057-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...