3,3'-Bipyridin-6,6'-dicarbonitril

Lieferant: COMBI-BLOCKS

PY-7880-100M
COBBPY-7880-100MEA 404 EUR
COBBPY-7880-100M
3,3'-Bipyridin-6,6'-dicarbonitril
3,3'-Bipyridin-6,6'-dicarbonitril
Formel: C₁₂H₆N₄
MDL: MFCD11855996
CAS-Nummer: 1133116-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR