5-Brom-1H-indazol

Lieferant: COMBI-BLOCKS

PY-7893-1G PY-7893-25G PY-7893-5G
COBBPY-7893-1GEA 0 EUR
COBBPY-7893-1G COBBPY-7893-25G COBBPY-7893-5G
5-Brom-1H-indazol
5-Bromo-1H-indazole
Formel: C₇H₅BrN₂
MDL: MFCD00839493
CAS-Nummer: 53857-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR