2-(Butylamino)-5-brompyridin

Lieferant: COMBI-BLOCKS

PY-7993-1G PY-7993-5G
COBBPY-7993-1GEA 0 EUR
COBBPY-7993-1G COBBPY-7993-5G
2-(Butylamino)-5-brompyridin
2-(Butylamino)-5-brompyridin
Formel: C₉H₁₃BrN₂
MDL: MFCD11123176
CAS-Nummer: 280116-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR