3-Brom-2-methylanisol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Bromo-3-methoxy-2-methylbenzene
QA-0118-1G
COBBQA-0118-1GEA
109
EUR
COBBQA-0118-1G
3-Brom-2-methylanisol
3-Brom-2-methylanisol
Formel:
C₈H₉BrO Molecular Weight: 201,0605 g/mol |
MDL:
MFCD11100991 CAS-Nummer: 31804-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...