4'-Brom-3'-fluoracetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-3-fluoro--acetophenone

QA-2571-1G
COBBQA-2571-1GEA 109 EUR
COBBQA-2571-1G
4'-Brom-3'-fluoracetophenon
4'-Brom-3'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217,04 g/mol
MDL: MFCD00466241
CAS-Nummer: 304445-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR