Isochinolin-3-ylmethanol

Lieferant: COMBI-BLOCKS

QA-2946-1G
COBBQA-2946-1GEA 235 EUR
COBBQA-2946-1G
Isochinolin-3-ylmethanol
Isochinolin-3-ylmethanol
Formel: C₁₀H₉NO
Molecular Weight: 159,1846 g/mol
MDL: MFCD01859798
CAS-Nummer: 76884-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR