N-Boc-D-alaninol

Lieferant: COMBI-BLOCKS

Synonyme: Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol

QA-4677-5G
COBBQA-4677-5GEA 0 EUR
COBBQA-4677-5G
N-Boc-D-alaninol
N-Boc-D-alaninol
Formel: C₈H₁₇NO₃
Molecular Weight: 175.2 g/mol
MDL: MFCD00235912
CAS-Nummer: 106391-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR