Brom-tris-pyrrolidinophosphoniumhexafluorophosphat

Lieferant: COMBI-BLOCKS

Synonyme: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

QA-4827-25G
COBBQA-4827-25GEA 162 EUR
COBBQA-4827-25G
Brom-tris-pyrrolidinophosphoniumhexafluorophosphat
Brom-tris-pyrrolidinophosphoniumhexafluorophosphat
Formel: C₁₂H₂₄BrF₆N₃P₂
Molecular Weight: 466,2 g/mol
MDL: MFCD00077412
CAS-Nummer: 132705-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR