2-[(4-Brombenzyl)oxy]essigsäure
Lieferant: COMBI-BLOCKS
QA-4837-1G
COBBQA-4837-1GEA
692
EUR
COBBQA-4837-1G
2-[(4-Brombenzyl)oxy]essigsäure
2-[(4-Brombenzyl)oxy]essigsäure
Formel:
C₉H₉BrO₃ |
MDL:
MFCD04123920 CAS-Nummer: 82499-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...