4-(Aminomethyl)tetrahydropyran-4-ol

Lieferant: COMBI-BLOCKS

QA-5766-1G
COBBQA-5766-1GEA 434 EUR
COBBQA-5766-1G
4-(Aminomethyl)tetrahydropyran-4-ol
4-(Aminomethyl)tetrahydropyran-4-ol
Formel: C₆H₁₃NO₂
MDL: MFCD01628294
CAS-Nummer: 783303-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR