3,5-Dichlorpyridazin

Lieferant: COMBI-BLOCKS

QA-6462-5G
COBBQA-6462-5GEA 336 EUR
COBBQA-6462-5G
3,5-Dichlorpyridazin
3,5-Dichloropyridazine
Formel: C₄H₂Cl₂N₂
Molecular Weight: 148.9781 g/mol
MDL: MFCD10698048
CAS-Nummer: 1837-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR