2-Amino-5-methoxypyrazin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methoxypyrazin-2-amine

QA-8419-1G
COBBQA-8419-1GEA 174 EUR
COBBQA-8419-1G
2-Amino-5-methoxypyrazin
2-Amino-5-methoxypyrazine
Formel: C₅H₇N₃O
Molecular Weight: 125,13 g/mol
MDL: MFCD09909647
CAS-Nummer: 54013-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR