4-Benzyloxy-2-hydroxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Benzyloxy-2(1H)-pyridone

QA-8863-25G
COBBQA-8863-25GEA 914 EUR
COBBQA-8863-25G
4-Benzyloxy-2-hydroxypyridin
4-Benzyloxy-2-hydroxypyridin
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201,22 g/mol
MDL: MFCD00191783
CAS-Nummer: 53937-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR