2-Chlor-6-nitrochinolin

Lieferant: COMBI-BLOCKS

QB-5686-1G
COBBQB-5686-1GEA 298 EUR
COBBQB-5686-1G
2-Chlor-6-nitrochinolin
2-Chloro-6-nitroquinoline
Formel: C₉H₅ClN₂O₂
Molecular Weight: 208,6000 g/mol
MDL: MFCD09040696
CAS-Nummer: 29969-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR