ω,3-Dibrom-4-methoxyacetophenon

Lieferant: COMBI-BLOCKS

QB-7514-1G
COBBQB-7514-1GEA 0 EUR
COBBQB-7514-1G
ω,3-Dibrom-4-methoxyacetophenon
ω,3-Dibrom-4-methoxyacetophenon
Formel: C₉H₈Br₂O₂
MDL: MFCD02646974
CAS-Nummer: 6096-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR