trans-3-Aminocyclopentanol Hydrochlorid

Lieferant: COMBI-BLOCKS

QB-8475-250MG
COBBQB-8475-250MGEA 0 EUR
COBBQB-8475-250MG
trans-3-Aminocyclopentanol Hydrochlorid
trans-3-Aminocyclopentanol Hydrochlorid
Formel: C₅H₁₂ClNO
MDL: MFCD22380009
CAS-Nummer: 124555-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR