4-(Isopropylsulphonyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Propane-2-sulphonyl)aniline

SH-5490-250MG
COBBSH-5490-250MGEA 0 EUR
COBBSH-5490-250MG
4-(Isopropylsulphonyl)anilin
4-(Isopropylsulphonyl)anilin
Formel: C₉H₁₃NO₂S
MDL: MFCD01566617
CAS-Nummer: 25355-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR