2-Amino-5-bromchinazolin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromoquinazolin-2-amine
SH-5674-1G
COBBSH-5674-1GEA
242
EUR
COBBSH-5674-1G
2-Amino-5-bromchinazolin
2-Amino-5-bromoquinazoline
Formel:
C₈H₆BrN₃ |
MDL:
MFCD09834658 CAS-Nummer: 181871-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...