3-Amino-5-fluor-1-methyl-1H-indazol
Lieferant: COMBI-BLOCKS
SS-0061-5G
COBBSS-0061-5GEA
0
EUR
COBBSS-0061-5G
3-Amino-5-fluor-1-methyl-1H-indazol
3-Amino-5-fluoro-1-methyl-1H-indazole
Formel:
C₈H₈FN₃ |
MDL:
MFCD09054760 CAS-Nummer: 171809-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...