3-Amino-6-fluor-1-methyl-1H-indazol

Lieferant: COMBI-BLOCKS

SS-0062-5G
COBBSS-0062-5GEA 0 EUR
COBBSS-0062-5G
3-Amino-6-fluor-1-methyl-1H-indazol
3-Amino-6-fluoro-1-methyl-1H-indazole
Formel: C₈H₈FN₃
MDL: MFCD11109398
CAS-Nummer: 171809-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR