(R)-(1-Boc-piperidin-3-yl)essigsäure

Lieferant: COMBI-BLOCKS

SS-1500-250MG
COBBSS-1500-250MGEA 0 EUR
COBBSS-1500-250MG
(R)-(1-Boc-piperidin-3-yl)essigsäure
(R)-(1-Boc-piperidin-3-yl)essigsäure
Formel: C₁₂H₂₁NO₄
MDL: MFCD03094721
CAS-Nummer: 912940-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR