2,4-Dichlor-L-phenylalanin

Lieferant: COMBI-BLOCKS

Synonyme: H-Phe(2,4-DiCl)-OH

SS-2911-1G
COBBSS-2911-1GEA 34.4 EUR
COBBSS-2911-1G
2,4-Dichlor-L-phenylalanin
2,4-Dichlor-L-phenylalanin
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234.1 g/mol
MDL: MFCD01860882
CAS-Nummer: 111119-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR