2-Fluor-3-iodpyridin

Lieferant: COMBI-BLOCKS
Achtung

SS-3009-1G SS-3009-25G SS-3009-5G
COBBSS-3009-1GEA 22 EUR
COBBSS-3009-1G COBBSS-3009-25G COBBSS-3009-5G
2-Fluor-3-iodpyridin
2-Fluor-3-iodpyridin
Formel: C₅H₃FIN
Molecular Weight: 222.99 g/mol
MDL: MFCD03095287
CAS-Nummer: 113975-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR