2-Fluor-3-iodpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
SS-3009-1G
SS-3009-25G
SS-3009-5G
COBBSS-3009-1GEA
22
EUR
COBBSS-3009-1G
COBBSS-3009-25G
COBBSS-3009-5G
2-Fluor-3-iodpyridin
2-Fluor-3-iodpyridin
Formel:
C₅H₃FIN Molecular Weight: 222.99 g/mol |
MDL:
MFCD03095287 CAS-Nummer: 113975-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...