Ethyl-2-amino-2-cyanoacetat-mono(4-methylbenzolsulfonat)

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-amino-2-cyanoacetate 4-methylbenzenesulphonate

SS-3239-1G
COBBSS-3239-1GEA 126 EUR
COBBSS-3239-1G
Ethyl-2-amino-2-cyanoacetat-mono(4-methylbenzolsulfonat)
Ethyl-2-amino-2-cyanoacetat-mono(4-methylbenzolsulfonat)
Formel: C₁₂H₁₆N₂O₅S
Molecular Weight: 300,3308 g/mol
MDL: MFCD00666813
CAS-Nummer: 37842-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR