4-Fluor-3-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Fluoro-4-methoxy-2-nitrobenzene

SS-3741-1G
COBBSS-3741-1GEA 0 EUR
COBBSS-3741-1G
4-Fluor-3-nitroanisol
4-Fluor-3-nitroanisol
Formel: C₇H₆FNO₃
MDL: MFCD11848442
CAS-Nummer: 61324-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR