2-Brom-4,5-dichlorimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Brom-4,5-dichlor-1H-imidazol
SS-4045-25G
COBBSS-4045-25GEA
808
EUR
COBBSS-4045-25G
2-Brom-4,5-dichlorimidazol
2-Bromo-4,5-dichloroimidazole
Formel:
C₃HBrCl₂N₂ Molecular Weight: 215,86 g/mol |
MDL:
MFCD00067894 CAS-Nummer: 16076-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...