α,3-Dibrom-4-fluortoluol

Lieferant: COMBI-BLOCKS

SS-4252-25G
COBBSS-4252-25GEA 0 EUR
COBBSS-4252-25G
α,3-Dibrom-4-fluortoluol
α,3-Dibrom-4-fluortoluol
Formel: C₇H₅Br₂F
MDL: MFCD09037367
CAS-Nummer: 78239-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR